Product Details

SNP ID
rs6088525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34553468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTTGGGGCCCTCCTCCTCCACG[C/T]TGGCCTCAGTTCAGGGCTGAGTCTG
Phenotype
MIM: 601242 MIM: 608528
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3A PubMed Links

Gene Details

Gene
MAP1LC3A
Gene Name
microtubule associated protein 1 light chain 3 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032514.3 Intron NP_115903.1
NM_181509.2 Intron NP_852610.1
XM_011529083.2 Intron XP_011527385.1
XM_011529084.2 Intron XP_011527386.1
XM_011529085.2 Intron XP_011527387.1
Gene
PIGU
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
There are no transcripts associated with this gene.

View Full Product Details