Product Details

SNP ID
rs2113143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22213540 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGAGAGCTGCTCTTCAGGACTT[C/T]GTTAGTTTCCAATTGCTGCTGTAAC
Phenotype
MIM: 606968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF2K PubMed Links

Gene Details

Gene
EEF2K
Gene Name
eukaryotic elongation factor 2 kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013302.3 Intron NP_037434.1
XM_017023193.1 Intron XP_016878682.1
XM_017023194.1 Intron XP_016878683.1

View Full Product Details