Product Details

SNP ID
rs1320213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40191918 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGAAGATGTTTCAAATGCTATTA[C/T]CTGGTTAGGGAGGCTGGTTTTTTTT
Phenotype
MIM: 602037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
N4BP2 PubMed Links

Gene Details

Gene
N4BP2
Gene Name
NEDD4 binding protein 2
There are no transcripts associated with this gene.

Gene
RHOH
Gene Name
ras homolog family member H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278359.1 Intron NP_001265288.1
NM_001278360.1 Intron NP_001265289.1
NM_001278361.1 Intron NP_001265290.1
NM_001278362.1 Intron NP_001265291.1
NM_001278363.1 Intron NP_001265292.1
NM_001278364.1 Intron NP_001265293.1
NM_001278365.1 Intron NP_001265294.1
NM_001278366.1 Intron NP_001265295.1
NM_001278367.1 Intron NP_001265296.1
NM_001278368.1 Intron NP_001265297.1
NM_001278369.1 Intron NP_001265298.1
NM_004310.4 Intron NP_004301.1
XM_011513692.1 Intron XP_011511994.1
XM_017008188.1 Intron XP_016863677.1
XM_017008189.1 Intron XP_016863678.1

View Full Product Details