Product Details

SNP ID
rs11757
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:171784535 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGATGTCCCTGTTTCCATATGCACA[C/G]TGACAGGCGAGGAGCTGCCTAGGAA
Phenotype
MIM: 603667
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105373738 PubMed Links

Gene Details

Gene
LOC105373738
Gene Name
uncharacterized LOC105373738
There are no transcripts associated with this gene.

Gene
SLC25A12
Gene Name
solute carrier family 25 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003705.4 2940 UTR 3 NP_003696.2
XM_011512070.2 2940 UTR 3 XP_011510372.1

View Full Product Details