Product Details
- SNP ID
-
rs555316
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:166604016 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGAACTTTCTAATAATCAGAGCTT[A/T]TTGAAAATGAATATACTGTCTATGA
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FMO9P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115547854] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FMO9P
- Gene Name
- flavin containing monooxygenase 9 pseudogene
There are no transcripts associated with this gene.
View Full Product Details