Product Details

SNP ID
rs3119810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34183249 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTATCAAAACTACGTGAGAGGGA[C/G]TTTTAAAAAATACATAAACCCGGCC
Phenotype
MIM: 610677
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LSM14A PubMed Links

Gene Details

Gene
LSM14A
Gene Name
LSM14A, mRNA processing body assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114093.1 Intron NP_001107565.1
NM_015578.2 Intron NP_056393.2
XM_005258719.4 Intron XP_005258776.1
XM_005258720.4 Intron XP_005258777.1
XM_005258721.4 Intron XP_005258778.1
XM_011526708.2 Intron XP_011525010.1
XM_017026575.1 Intron XP_016882064.1
XM_017026576.1 Intron XP_016882065.1
XM_017026577.1 Intron XP_016882066.1

View Full Product Details