Product Details

SNP ID
rs9285530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151251359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTATAAGGCAGAGATAACATCC[G/T]CACAGTAAAGTGGTGGGAAGTGTGA
Phenotype
MIM: 604698
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP12 PubMed Links

Gene Details

Gene
AKAP12
Gene Name
A-kinase anchoring protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005100.3 Intron NP_005091.2
NM_144497.2 Intron NP_653080.1
XM_005267235.2 Intron XP_005267292.1
XM_017011517.1 Intron XP_016867006.1
XM_017011518.1 Intron XP_016867007.1

View Full Product Details