Product Details

SNP ID
rs839761
Assay Type
Validated
NCBI dbSNP Submissions
37
Location
Chr.1:43398186 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAGTAGTGTTGACACTAGTGTTAAG[A/G]TGATCTCATCTTGAGAAAAGTTGTA
Phenotype
MIM: 607956 MIM: 615463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED8 PubMed Links

Gene Details

Gene
MED8
Gene Name
mediator complex subunit 8
There are no transcripts associated with this gene.

Gene
SZT2
Gene Name
seizure threshold 2 homolog (mouse)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015284.3 Intron NP_056099.3
XM_005270686.3 Intron XP_005270743.1
XM_006710501.3 Intron XP_006710564.1
XM_011541106.2 Intron XP_011539408.1
XM_011541107.2 Intron XP_011539409.1
XM_017000819.1 Intron XP_016856308.1
XM_017000820.1 Intron XP_016856309.1
XM_017000821.1 Intron XP_016856310.1

View Full Product Details