Product Details
- SNP ID
-
rs11587687
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:109605179 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTGGGTGAAATTAACATTTTTCAC[A/G]CCTCTAACCTTTCCCAAATTCAGAG
- Phenotype
-
MIM: 139370
MIM: 139340
MIM: 611189
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNAI3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111403662] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNAI3
- Gene Name
- G protein subunit alpha i3
There are no transcripts associated with this gene.
- Gene
- GNAT2
- Gene Name
- G protein subunit alpha transducin 2
- Gene
- MIR197
- Gene Name
- microRNA 197
There are no transcripts associated with this gene.
View Full Product Details