Product Details

SNP ID
rs3824473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101572495 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCTGCTGCCTGTCTTCTGTTTC[C/T]GCTTACCACATCCCCTCCCCTTTTC
Phenotype
MIM: 606650 MIM: 607699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIN3A PubMed Links

Gene Details

Gene
GRIN3A
Gene Name
glutamate ionotropic receptor NMDA type subunit 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133445.2 4628 UTR 3 NP_597702.2
XM_011518211.2 4628 Intron XP_011516513.1
Gene
RNF20
Gene Name
ring finger protein 20
There are no transcripts associated with this gene.

View Full Product Details