Product Details

SNP ID
rs169082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:169647052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCAGAGAGCACCATTATTAGTA[C/T]TAATGTCCCCATTGGACAGGTTGGG
Phenotype
MIM: 603122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK2 PubMed Links

Gene Details

Gene
DOCK2
Gene Name
dedicator of cytokinesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004946.2 Intron NP_004937.1
XM_005265830.3 Intron XP_005265887.1
XM_011534448.2 Intron XP_011532750.1
XM_011534449.2 Intron XP_011532751.1
XM_011534450.2 Intron XP_011532752.1
XM_011534451.2 Intron XP_011532753.1
XM_017009189.1 Intron XP_016864678.1
XM_017009190.1 Intron XP_016864679.1

View Full Product Details