Product Details

SNP ID
rs34769775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80696831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAACAAGCAAACCAAAAACCTGA[C/T]TTTTATTCACTATTAGACAAAAGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD17C PubMed Links

Gene Details

Gene
ABHD17C
Gene Name
abhydrolase domain containing 17C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021214.1 Intron NP_067037.1

View Full Product Details