Product Details

SNP ID
rs908702
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:206932296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAGCAGCTCTGAATTTCAGTTT[C/T]GAGTATGTGTGGGTTTATCTTTCCC
Phenotype
MIM: 173880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGR PubMed Links

Gene Details

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 Intron NP_002635.2
XM_011509629.1 Intron XP_011507931.1

View Full Product Details