Product Details

SNP ID
rs11739838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150592605 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTCCATGCAGGAGAAGGAGGCA[C/T]TGGGGCTGGACCCATAGGCCAGCTA
Phenotype
MIM: 608155
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SYNPO PubMed Links

Gene Details

Gene
SYNPO
Gene Name
synaptopodin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109974.2 Intron NP_001103444.1
NM_001166208.1 Intron NP_001159680.1
NM_001166209.1 Intron NP_001159681.1
NM_007286.5 Intron NP_009217.3
XM_005268370.1 Intron XP_005268427.1
XM_005268371.1 Intron XP_005268428.1
XM_006714755.3 Intron XP_006714818.1
XM_011537552.2 Intron XP_011535854.1
XM_017009007.1 Intron XP_016864496.1
XM_017009008.1 Intron XP_016864497.1

View Full Product Details