Product Details

SNP ID
rs4849073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112543081 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTTCAAGGTTATCCTTCAGTTT[C/G]CTTCTCTTAGAGCTTTAAAGAGAGA
Phenotype
MIM: 602000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105373562 PubMed Links

Gene Details

Gene
LOC105373562
Gene Name
uncharacterized LOC105373562
There are no transcripts associated with this gene.

Gene
POLR1B
Gene Name
RNA polymerase I subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137604.2 Intron NP_001131076.1
NM_001282772.1 Intron NP_001269701.1
NM_001282774.1 Intron NP_001269703.1
NM_001282776.1 Intron NP_001269705.1
NM_001282777.1 Intron NP_001269706.1
NM_001282779.1 Intron NP_001269708.1
NM_019014.5 Intron NP_061887.2

View Full Product Details