Product Details
- SNP ID
-
rs11123139
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112547573 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACCATGAGGAGGCAGAGGTAAT[C/G]ACGGGCGTCCAGGCATGAGACAGTA
- Phenotype
-
MIM: 602000
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105373562
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78234807] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105373562
- Gene Name
- uncharacterized LOC105373562
There are no transcripts associated with this gene.
- Gene
- POLR1B
- Gene Name
- RNA polymerase I subunit B
View Full Product Details