Product Details

SNP ID
rs2044343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68529021 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAACGGTGCAGACTAATCCTAAAT[C/T]AGTCTTTCTCTTTGCCTCAACAACT
Phenotype
MIM: 605002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO2B PubMed Links

Gene Details

Gene
CORO2B
Gene Name
coronin 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190456.1 Intron NP_001177385.1
NM_001190457.1 Intron NP_001177386.1
NM_001324014.1 Intron NP_001310943.1
NM_001324015.1 Intron NP_001310944.1
NM_006091.4 Intron NP_006082.3

View Full Product Details