Product Details

SNP ID
rs4725570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142244463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCAAATTATCTGTCTAAAGAAA[A/G]AAAGTCTCCACATTTGCCACATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOXD2P PubMed Links

Gene Details

Gene
MOXD2P
Gene Name
monooxygenase, DBH-like 2, pseudogene
There are no transcripts associated with this gene.

Gene
PRSS58
Gene Name
protease, serine 58
There are no transcripts associated with this gene.

View Full Product Details