Product Details

SNP ID
rs736479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25880756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTCTCCCCACCCAAAATTACTT[C/T]GATTATTTAATGTCTGTAGTCATCC
Phenotype
MIM: 104760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APP PubMed Links

Gene Details

Gene
APP
Gene Name
amyloid beta precursor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000484.3 3316 UTR 3 NP_000475.1
NM_001136016.3 3316 UTR 3 NP_001129488.1
NM_001136129.2 3316 UTR 3 NP_001129601.1
NM_001136130.2 3316 UTR 3 NP_001129602.1
NM_001136131.2 3316 UTR 3 NP_001129603.1
NM_001204301.1 3316 UTR 3 NP_001191230.1
NM_001204302.1 3316 UTR 3 NP_001191231.1
NM_001204303.1 3316 UTR 3 NP_001191232.1
NM_201413.2 3316 UTR 3 NP_958816.1
NM_201414.2 3316 UTR 3 NP_958817.1

View Full Product Details