Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039670.2 | 2574 | UTR 3 | NP_001034759.1 | ||
NM_001193457.1 | 2574 | UTR 3 | NP_001180386.1 | ||
NM_080730.4 | 2574 | UTR 3 | NP_542768.2 | ||
XM_006718975.3 | 2574 | UTR 3 | XP_006719038.1 | ||
XM_006718976.3 | 2574 | UTR 3 | XP_006719039.1 | ||
XM_006718977.3 | 2574 | UTR 3 | XP_006719040.1 | ||
XM_006718981.3 | 2574 | UTR 3 | XP_006719044.1 | ||
XM_011520947.2 | 2574 | UTR 3 | XP_011519249.1 | ||
XM_011520948.2 | 2574 | UTR 3 | XP_011519250.1 | ||
XM_011520949.2 | 2574 | UTR 3 | XP_011519251.1 | ||
XM_011520952.2 | 2574 | UTR 3 | XP_011519254.1 | ||
XM_017019169.1 | 2574 | Intron | XP_016874658.1 | ||
XM_017019170.1 | 2574 | UTR 3 | XP_016874659.1 | ||
XM_017019171.1 | 2574 | UTR 3 | XP_016874660.1 | ||
XM_017019172.1 | 2574 | UTR 3 | XP_016874661.1 | ||
XM_017019173.1 | 2574 | UTR 3 | XP_016874662.1 |