Product Details
- SNP ID
-
rs238648
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23522883 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGTGTTTGGCGTGAACCATTGCAG[A/G]GATTACTTATTAGTTACAAGGGGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST13P
PubMed Links
Gene Details
- Gene
- CST13P
- Gene Name
- cystatin 13, pseudogene
There are no transcripts associated with this gene.
View Full Product Details