Product Details

SNP ID
rs104894774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136648355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTGTTTTTCTTATCACCCAGA[G/T]GATTGGGTCAGCACTTTTTGCTGTG
Phenotype
MIM: 300386
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD40LG PubMed Links

Gene Details

Gene
CD40LG
Gene Name
CD40 ligand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000074.2 179 Missense Mutation AGG,ATG R36M NP_000065.1
Gene
LINC00892
Gene Name
long intergenic non-protein coding RNA 892
There are no transcripts associated with this gene.

View Full Product Details