Product Details

SNP ID
rs33675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36154351 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGTGCATAAATGTGACGTATTT[A/G]TTCCAGGAACAGGGAAGCCATATTC
Phenotype
MIM: 601436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMBRD2 PubMed Links

Gene Details

Gene
LMBRD2
Gene Name
LMBR1 domain containing 2
There are no transcripts associated with this gene.

Gene
MIR580
Gene Name
microRNA 580
There are no transcripts associated with this gene.

Gene
SKP2
Gene Name
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243120.1 Intron NP_001230049.1
NM_005983.3 Intron NP_005974.2
NM_032637.3 Intron NP_116026.1
XM_011514081.2 Intron XP_011512383.1
XM_011514082.2 Intron XP_011512384.1
XM_011514083.2 Intron XP_011512385.1
XM_017009753.1 Intron XP_016865242.1

View Full Product Details