Product Details

SNP ID
rs1075972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3113967 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGAGTTGGAGGAGTATGAGTCA[G/T]CGACAGGCCAGGGGCCAGTCCCTGG
Phenotype
MIM: 603436
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF205 PubMed Links

Gene Details

Gene
ZNF205
Gene Name
zinc finger protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042428.1 Intron NP_001035893.1
NM_001278158.1 Intron NP_001265087.1
NM_003456.2 Intron NP_003447.2
XM_005255558.2 Intron XP_005255615.1
Gene
ZNF205-AS1
Gene Name
ZNF205 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details