Product Details
- SNP ID
-
rs369487
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:79203011 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATTTTTGTTTGTTTCTAGTCTAT[G/T]GGCATCAAGTGCTTTAAAATGCTCA
- Phenotype
-
MIM: 177075
MIM: 605131
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105371352
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77596276] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105371352
- Gene Name
- uncharacterized LOC105371352
There are no transcripts associated with this gene.
- Gene
- MAF
- Gene Name
- MAF bZIP transcription factor
- Gene
- WWOX
- Gene Name
- WW domain containing oxidoreductase
View Full Product Details