Product Details

SNP ID
rs448189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22820137 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTGGTCCAGGCGAACGTCACAC[A/G]GTATGATTAAATGAAACCATTGAGT
Phenotype
MIM: 604056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS3ST2 PubMed Links

Gene Details

Gene
HS3ST2
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006043.1 Intron NP_006034.1
XM_011546001.2 Intron XP_011544303.1
XM_011546002.2 Intron XP_011544304.1
Gene
LOC107984851
Gene Name
collagen alpha-1(XVI) chain-like
There are no transcripts associated with this gene.

View Full Product Details