Product Details

SNP ID
rs2032630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19704954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATCAGATTTAGGACACAAAAGC[T/A]ACTACATAATGAAAAAGAGAGCTGG
Phenotype
MIM: 426000
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 6126 Intron NP_001140177.1
NM_001146706.1 6126 Intron NP_001140178.1
NM_004653.4 6126 Intron NP_004644.2
XM_005262560.2 6126 UTR 3 XP_005262617.1
XM_005262561.2 6126 UTR 3 XP_005262618.1
XM_011531468.2 6126 UTR 3 XP_011529770.1

View Full Product Details