Product Details

SNP ID
rs3768571
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:22027834 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
CCTAAGAAATGGGTCAGGGAACCTT[C/T]AGAGAAGGGATGACTAGGTGCTGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00339 PubMed Links

Gene Details

Gene
LINC00339
Gene Name
long intergenic non-protein coding RNA 339
There are no transcripts associated with this gene.

Gene
LOC101928043
Gene Name
uncharacterized LOC101928043
There are no transcripts associated with this gene.

View Full Product Details