Product Details

SNP ID
rs556625
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:208022636 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTACACACGCAAGAATATCTGTT[C/T]TGTTTGGGGGGAGTATTTCCATTCT
Phenotype
MIM: 601054
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNA2 PubMed Links

Gene Details

Gene
PLXNA2
Gene Name
plexin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025179.3 11309 UTR 3 NP_079455.3
XM_005273164.2 11309 UTR 3 XP_005273221.1
XM_005273165.3 11309 Intron XP_005273222.1

View Full Product Details