Product Details

SNP ID
rs846898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44613466 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCCACGGCCCTGTCTGACTCTGG[A/G]GCAGGCACTATTATTAACGCCTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGSF23 PubMed Links

Gene Details

Gene
IGSF23
Gene Name
immunoglobulin superfamily member 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205280.1 890 Intron NP_001192209.1
XM_017026317.1 890 Intron XP_016881806.1
XM_017026318.1 890 UTR 5 XP_016881807.1
XM_017026319.1 890 Intron XP_016881808.1
XM_017026320.1 890 Intron XP_016881809.1
XM_017026321.1 890 Intron XP_016881810.1
XM_017026322.1 890 Intron XP_016881811.1
XM_017026323.1 890 Intron XP_016881812.1
XM_017026324.1 890 Intron XP_016881813.1
XM_017026325.1 890 Intron XP_016881814.1
XM_017026326.1 890 Intron XP_016881815.1

View Full Product Details