Product Details

SNP ID
rs12921264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2020432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCCCTTCCATTCCCTGCTCCGC[A/G]CCCAGAAGAGCTTTCCCTGGGCAGG
Phenotype
MIM: 607997 MIM: 606553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPW PubMed Links

Gene Details

Gene
NPW
Gene Name
neuropeptide W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099456.2 Intron NP_001092926.2
Gene
SLC9A3R2
Gene Name
SLC9A3 regulator 2
There are no transcripts associated with this gene.

View Full Product Details