Product Details

SNP ID
rs7966590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:461674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACTCTGACTCTGACCTGTCGTC[A/G]TGTCAGCTTCTCTACCCAGAGTTGG
Phenotype
MIM: 612220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT3 PubMed Links

Gene Details

Gene
B4GALNT3
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173593.3 Intron NP_775864.3

View Full Product Details