Product Details

SNP ID
rs611481
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:3708638 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCCTTCCCCAGCTGAGGGTCCTT[A/G]GACTCCCCCCCAACCTCCCCCACCA
Phenotype
MIM: 606102 MIM: 612689
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIP5K1C PubMed Links

Gene Details

Gene
PIP5K1C
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
There are no transcripts associated with this gene.

Gene
TJP3
Gene Name
tight junction protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267560.1 Intron NP_001254489.1
NM_001267561.1 Intron NP_001254490.1

View Full Product Details