Product Details

SNP ID
rs4762834
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:21504920 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATACTCCAGTGTCCTTGAGTTTCA[C/T]GTAACCAAACACCTTTAACCTAAAA
Phenotype
MIM: 615078 MIM: 600537
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOLT1B PubMed Links

Gene Details

Gene
GOLT1B
Gene Name
golgi transport 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016072.4 Intron NP_057156.1
XM_017019384.1 Intron XP_016874873.1
Gene
RECQL
Gene Name
RecQ like helicase
There are no transcripts associated with this gene.

View Full Product Details