Product Details

SNP ID
rs352494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4185617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGCCCAGGTGGACAGAAGTGCA[C/T]GTGTGCACACGCCCCACACTCAACG
Phenotype
MIM: 606211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD24 PubMed Links

Gene Details

Gene
ANKRD24
Gene Name
ankyrin repeat domain 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133475.1 Intron NP_597732.1
XM_011527756.2 Intron XP_011526058.1
XM_011527757.2 Intron XP_011526059.1
XM_011527758.2 Intron XP_011526060.1
XM_011527761.1 Intron XP_011526063.1
XM_011527764.2 Intron XP_011526066.1
XM_011527765.2 Intron XP_011526067.1
XM_011527768.2 Intron XP_011526070.1
XM_017026438.1 Intron XP_016881927.1
XM_017026439.1 Intron XP_016881928.1
XM_017026440.1 Intron XP_016881929.1
XM_017026441.1 Intron XP_016881930.1
Gene
SIRT6
Gene Name
sirtuin 6
There are no transcripts associated with this gene.

View Full Product Details