Product Details
- SNP ID
-
rs4367573
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:7039361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCTTCTGGGCTGGCATCTCAT[C/G]AGCTCTTGCCGGGAGCGGCTCTGCC
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DEFA11P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116956725] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEFA11P
- Gene Name
- defensin alpha 11, pseudogene
There are no transcripts associated with this gene.
- Gene
- DEFA7P
- Gene Name
- defensin alpha 7, pseudogene
There are no transcripts associated with this gene.
View Full Product Details