Product Details
- SNP ID
-
rs11704378
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:16787262 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCAAAGAACAAATGTATTAGTCAC[G/T]GGTGTTCAAGGAAAAGCTGGGTGCA
- Phenotype
-
MIM: 611674
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
XKR3
PubMed Links
Gene Details
- Gene
- XKR3
- Gene Name
- XK related 3
View Full Product Details