Product Details

SNP ID
rs7664
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:47216345 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TTCCCGATACATCCTCACATATATA[G/T]ACACATCATCACCACATGGGTTTTT
Phenotype
MIM: 187040
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAL1 PubMed Links

Gene Details

Gene
TAL1
Gene Name
TAL bHLH transcription factor 1, erythroid differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287347.2 4697 UTR 3 NP_001274276.1
NM_001290403.1 4697 UTR 3 NP_001277332.1
NM_001290404.1 4697 UTR 3 NP_001277333.1
NM_001290405.1 4697 UTR 3 NP_001277334.1
NM_001290406.1 4697 UTR 3 NP_001277335.1
NM_003189.5 4697 UTR 3 NP_003180.1
XM_005271160.4 4697 UTR 3 XP_005271217.1
XM_017002187.1 4697 UTR 3 XP_016857676.1
XM_017002188.1 4697 UTR 3 XP_016857677.1
XM_017002189.1 4697 UTR 3 XP_016857678.1
XM_017002190.1 4697 UTR 3 XP_016857679.1
XM_017002191.1 4697 UTR 3 XP_016857680.1
XM_017002192.1 4697 Intron XP_016857681.1
XM_017002193.1 4697 UTR 3 XP_016857682.1

View Full Product Details