Product Details

SNP ID
rs1572343
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:93150645 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTAAAATTTATCATTTGGAACTG[C/T]CTTTTCTTTCTGCAATGGTATTACG
Phenotype
MIM: 616876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 5211 UTR 3 NP_001161302.1
NM_016040.4 5211 UTR 3 NP_057124.3

View Full Product Details