Product Details

SNP ID
rs8192709
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:40991369 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
AGGACTCTTGCTACTCCTGGTTCAG[C/T]GCCACCCTAACACCCATGACCGCCT
Phenotype
MIM: 123930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
CYP2B6*10,c.64C>T CYP2B6*10,g.64C>T CYP2B6*2A,c.64C>T CYP2B6*2A,g.64C>T CYP2B6*2B,c.64C>T CYP2B6*2B,g.64C>T
Literature Links
CYP2B6 PubMed Links

Gene Details

Gene
CYP2B6
Gene Name
cytochrome P450 family 2 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000767.4 167 Missense Mutation CGC,TGC R22C NP_000758.1
XM_005258569.4 167 Missense Mutation CGC,TGC R22C XP_005258626.1
XM_006723050.3 167 Missense Mutation CGC,TGC R22C XP_006723113.1
XM_011526546.2 167 Missense Mutation CGC,TGC R22C XP_011524848.1
XM_011526547.2 167 Intron XP_011524849.1
XM_011526548.2 167 Missense Mutation CGC,TGC R22C XP_011524850.1
XM_011526549.2 167 Intron XP_011524851.1
XM_011526550.2 167 Intron XP_011524852.1

View Full Product Details