Product Details

SNP ID
rs2250122
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:152122333 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGAGGGCTAAAGCTGCCACACCGA[G/T]GGCTTTCGCCAAGATCAAGGTCCTC
Phenotype
MIM: 608441
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYNE1 PubMed Links

Gene Details

Gene
SYNE1
Gene Name
spectrin repeat containing nuclear envelope protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033071.3 26957 UTR 3 NP_149062.1
NM_182961.3 26957 UTR 3 NP_892006.3
XM_006715407.1 26957 UTR 3 XP_006715470.1
XM_006715408.2 26957 UTR 3 XP_006715471.1
XM_006715409.1 26957 UTR 3 XP_006715472.1
XM_006715410.2 26957 UTR 3 XP_006715473.1
XM_006715411.1 26957 UTR 3 XP_006715474.1
XM_006715412.2 26957 UTR 3 XP_006715475.1
XM_006715413.2 26957 UTR 3 XP_006715476.1
XM_006715414.1 26957 UTR 3 XP_006715477.1
XM_006715415.2 26957 UTR 3 XP_006715478.1
XM_006715416.2 26957 UTR 3 XP_006715479.1
XM_006715417.2 26957 UTR 3 XP_006715480.1
XM_006715420.2 26957 UTR 3 XP_006715483.1
XM_006715421.2 26957 UTR 3 XP_006715484.1
XM_006715422.1 26957 UTR 3 XP_006715485.1
XM_006715423.2 26957 UTR 3 XP_006715486.1
XM_006715424.2 26957 UTR 3 XP_006715487.1
XM_006715425.2 26957 UTR 3 XP_006715488.1
XM_011535641.2 26957 UTR 3 XP_011533943.1
XM_011535642.2 26957 UTR 3 XP_011533944.1
XM_011535643.1 26957 UTR 3 XP_011533945.1
XM_011535644.1 26957 UTR 3 XP_011533946.1
XM_011535645.2 26957 UTR 3 XP_011533947.1
XM_017010608.1 26957 UTR 3 XP_016866097.1
XM_017010609.1 26957 UTR 3 XP_016866098.1
XM_017010610.1 26957 UTR 3 XP_016866099.1
XM_017010611.1 26957 UTR 3 XP_016866100.1
XM_017010612.1 26957 UTR 3 XP_016866101.1
XM_017010613.1 26957 UTR 3 XP_016866102.1
XM_017010614.1 26957 UTR 3 XP_016866103.1
XM_017010615.1 26957 UTR 3 XP_016866104.1
XM_017010616.1 26957 UTR 3 XP_016866105.1
XM_017010617.1 26957 UTR 3 XP_016866106.1
XM_017010618.1 26957 UTR 3 XP_016866107.1
XM_017010619.1 26957 UTR 3 XP_016866108.1

View Full Product Details