Product Details

SNP ID
rs3749820
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:133197371 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTAGGCTTTTGAGGTGAGCTTCCTC[A/T]ACTTATATCTACATAGCATATCTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FSTL4 PubMed Links

Gene Details

Gene
FSTL4
Gene Name
follistatin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015082.1 3932 UTR 3 NP_055897.1
XM_011543283.1 3932 UTR 3 XP_011541585.1
XM_011543284.1 3932 UTR 3 XP_011541586.1
XM_011543286.2 3932 UTR 3 XP_011541588.1
XM_011543287.2 3932 UTR 3 XP_011541589.1
XM_017009251.1 3932 UTR 3 XP_016864740.1

View Full Product Details