Product Details

SNP ID
rs11211046
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:44855891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTTAGTAGGGAAGGCTGATTCA[C/T]GGATAATTGCCATGGCTTGTGAAAC
Phenotype
MIM: 606273
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2B3 PubMed Links

Gene Details

Gene
EIF2B3
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166588.2 Intron NP_001160060.1
NM_001261418.1 Intron NP_001248347.1
NM_020365.4 Intron NP_065098.1
XM_017002745.1 Intron XP_016858234.1
XM_017002746.1 Intron XP_016858235.1
XM_017002747.1 Intron XP_016858236.1

View Full Product Details