Product Details
- SNP ID
-
rs227933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:23584951 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACATATTCACAGGTATATAGAGAA[C/G]CAGCAGGAAGGAGTGAACTCTGGCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLK2P1
PubMed Links
Gene Details
- Gene
- CLK2P1
- Gene Name
- CDC like kinase 2, pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details