Product Details

SNP ID
rs642898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111703429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCTTTATATAACTCATGTAACC[A/G]CAATAGCTCTACTAAAATATAATTT
Phenotype
MIM: 611181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAD10 PubMed Links

Gene Details

Gene
ACAD10
Gene Name
acyl-CoA dehydrogenase family member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136538.1 Intron NP_001130010.1
NM_025247.5 Intron NP_079523.3

View Full Product Details