Product Details

SNP ID
rs12485150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:43414839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAGTACCTTTGTCTGCCTGCCC[C/T]TCTGGAGCAAGGGTCCTGGGGCAAA
Phenotype
MIM: 602112
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPPED1 PubMed Links

Gene Details

Gene
MPPED1
Gene Name
metallophosphoesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044370.1 2425 Intron NP_001037835.1
XM_011530367.2 2425 UTR 5 XP_011528669.1

View Full Product Details