Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001320561.1 | Intron | NP_001307490.1 | |||
NM_001320564.1 | Intron | NP_001307493.1 | |||
NM_001320565.1 | Intron | NP_001307494.1 | |||
NM_001320566.1 | Intron | NP_001307495.1 | |||
NM_001320567.1 | Intron | NP_001307496.1 | |||
NM_001320568.1 | Intron | NP_001307497.1 | |||
NM_001320569.1 | Intron | NP_001307498.1 | |||
NM_030818.3 | Intron | NP_110445.1 | |||
XM_005260086.4 | Intron | XP_005260143.1 | |||
XM_011528326.1 | Intron | XP_011526628.1 |