Product Details

SNP ID
rs41290504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:99533278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGCGCCCTGCATGCTGGCCGC[A/C]GCTGAGGGGACGCAATTTTCTGACG
Phenotype
MIM: 606727
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01475 PubMed Links

Gene Details

Gene
LINC01475
Gene Name
long intergenic non-protein coding RNA 1475
There are no transcripts associated with this gene.

Gene
NKX2-3
Gene Name
NK2 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145285.2 346 Silent Mutation GCA,GCC A49A NP_660328.2
XM_011539370.1 346 Silent Mutation GCA,GCC A49A XP_011537672.1

View Full Product Details