Product Details

SNP ID
rs211526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:496116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAGAGAGAAAAAAATCACCAAA[A/G]GTATATTTTCTTAGTAGAACCAAGT
Phenotype
MIM: 612793 MIM: 173320
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTDSS2 PubMed Links

Gene Details

Gene
PTDSS2
Gene Name
phosphatidylserine synthase 2
There are no transcripts associated with this gene.

Gene
RNH1
Gene Name
ribonuclease/angiogenin inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002939.3 Intron NP_002930.2
NM_203383.1 Intron NP_976317.1
NM_203384.1 Intron NP_976318.1
NM_203385.1 Intron NP_976319.1
NM_203386.2 Intron NP_976320.1
NM_203387.2 Intron NP_976321.1
NM_203388.2 Intron NP_976322.1
NM_203389.2 Intron NP_976323.1
XM_011520255.1 Intron XP_011518557.1
XM_011520257.2 Intron XP_011518559.1
XM_011520258.2 Intron XP_011518560.1
XM_011520259.2 Intron XP_011518561.1
XM_011520260.2 Intron XP_011518562.1
XM_011520261.2 Intron XP_011518563.1
XM_011520262.2 Intron XP_011518564.1
XM_011520263.1 Intron XP_011518565.1
XM_017018106.1 Intron XP_016873595.1

View Full Product Details