Product Details
- SNP ID
-
rs11800513
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:111724732 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACAAAGGATTTGCAGTGGCTGG[C/T]TCCACCAGAGTGGGCTTGCTCAGTT
- Phenotype
-
MIM: 179520
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM212B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78431345] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM212B
- Gene Name
- family with sequence similarity 212 member B
- Gene
- RAP1A
- Gene Name
- RAP1A, member of RAS oncogene family
There are no transcripts associated with this gene.
View Full Product Details